Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H,1H,2H,2H-Perfluorooctanesulfonic acid 25% in aqueous solution

Supplier: Matrix Scientific
Synonyms: 1H,1H,2H,2H-Tridecafluorooctane-1-sulphonic acid

009120-1G
101779-882EA 512.31 USD InStock
101779-882
1H,1H,2H,2H-Perfluorooctanesulfonic acid 25% in aqueous solution
1H,1H,2H,2H-Perfluorooctanesulfonic acid
Hazard: Corrosive/Irritant


Formula: C₈H₅F₁₃O₃S
MW: 428.17 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00042455
CAS Number: 27619-97-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty