Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

indane-5-sulfonyl chloride

Supplier: Matrix Scientific
Synonyms: 2,3-Dihydro-1H-indene-5-sulfonyl chloride2,3-Dihydro-1H-indene-5-sulphonyl chloride

011907-1G
101780-098EA 215.38 USD InStock
101780-098
indane-5-sulfonyl chloride
indane-5-sulfonyl chloride
Hazard: Corrosive/Moisture Sensitive


Formula: C₉H₉ClO₂S
MW: 216.679 g/mol
Boiling Pt: 180 °C (16 Torr)
Melting Pt: 46 °C
MDL Number: MFCD05237217
CAS Number: 52205-85-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty