Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Bromo-2,3,5,6-tetrafluorotoluene ≥97%

Supplier: Matrix Scientific
Synonyms: 2,3,5,6-Tetrafluorobenzylbromide2,3,5,6-Tetrafluorobenzyl bromide

012890-5G
101780-160EA 423.08 USD InStock
101780-160
α-Bromo-2,3,5,6-tetrafluorotoluene ≥97%
α-Bromo-2,3,5,6-tetrafluorotoluene
Hazard: Corrosive/Lachrymator


Formula: C₇H₃BrF₄
MW: 243 g/mol
Boiling Pt: 79…80 °C (80 mmHg)
MDL Number: MFCD00013549
CAS Number: 53001-73-3
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Boiling point 79-80°/80m
ND20 1.4885

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty