Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dimethylphenyl isocyanate

Supplier: Matrix Scientific
Synonyms: 3,5-Dimethylphenylisocyanate3,5-Xylyl isocyanate

038308-5G 038308-25G
101780-598EA 106.15 USD InStock
101780-598 101780-600
3,5-Dimethylphenyl isocyanate
3,5-Dimethylphenyl isocyanate
Hazard: Toxic


Formula: C₉H₉NO
MW: 147.18 g/mol
MDL Number: MFCD00013868
CAS Number: 54132-75-1
UN: 2206
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty