Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichlorobenzylamine

Supplier: Matrix Scientific

057004-5G 057004-1G
101780-748EA 21.54 USD InStock
101780-748 101780-750
3,5-Dichlorobenzylamine
3,5-Dichlorobenzylamine
Hazard: Corrosive


Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 141…144 °C (15 mmHg)
MDL Number: MFCD00052681
CAS Number: 39989-43-0
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Boiling point 141-144°/1

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty