Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Color filters

Supplier: EISCO SCIENTIFIC LLC
PH0648
470137-544PK 14.2 USD InStock
470137-544
Color filters
Educational Materials Physics Educational Materials Optics Learning Activities
Lightweight and Abrasion-Resistant

A set of three primary and three secondary acetate filters suitable for use in optical boxes and more. Mounted in plastic frames measuring 50mm x 50mm. The primary colors are: red, deep blue, dark green; and the secondary colors are: magenta, yellow, peacock blue.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty