Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Astrocyte Mouse Monoclonal Antibody [clone: ASTRO1 (10E4/R5)]

Supplier: GeneTex
GTX23268
89351-952EA 608.7 USD InStock
89351-952
Anti-Astrocyte Mouse Monoclonal Antibody [clone: ASTRO1 (10E4/R5)]
Antibodies
Astrocytes are the most plentiful and diverse of the glial cells. It appears that they play an important role in guiding the construction of the nervous system and controlling the chemical and ionic environment of neurons.

Type: Primary
Antigen: Astrocyte
Clonality: Monoclonal
Clone: [ASTRO1 (10E4/R5)]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty