Would you like to log in again?
Stirrer Magnets, Octagonal, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13654-22 13654-14 13654-24 13654-20 13654-06 13654-18 13654-36 13654-30 13654-02 13654-46 13654-32 13654-04 13654-48 13654-10 13654-38 13654-16 13654-28 13654-08 13654-12
89077-858EA 15.91 USD InStock
89077-858 89077-836 89077-846 89077-868 89077-838 89077-848 89077-832 89077-842 89077-864 89077-834 89077-856 89077-866 89077-844 89077-850 89077-860 89077-852 89077-830 89077-840 89077-870
Stirrer Magnets, Octagonal, PTFE, Ace Glass Incorporated
Stirrers Magnetic Stirring Bars
These octagonal PTFE stirrer magnets feature a center pivot ring and are available in a wide variety of lengths and diameters
They are ideally suited for use in containers with bases that are slightly curved or uneven.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty