Would you like to log in again?
SP Bel-Art Dispensing Jug, 5-Gallon, HDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art
H11850-0000
70690-038CS 261.04 USD InStock
70690-038
SP Bel-Art Dispensing Jug, 5-Gallon, HDPE, Bel-Art Products, a part of SP
Bottles Carboys
Use this 20 liters (5 gallon) jug with molded-in gradations (in gallons) to store and dispense common solutions.
  • Jug is made from strong and lightweight HDPE
  • LDPE spigot features a long stem for dispensing
  • Built-in handle provides for easy transport
This can used for common solutions including janitorial cleaners, deionized/distilled water, or other aqueous liquids.
Dimensions: 25 sq.×38 high cm( (9⁷/₈×15").
Ordering information: Optional 5-shelf dispensing rack is sold seperately, and does not include jugs or bottles.
Packaging: Shipping weight 1.7 kg (3.7 lbs.).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty