Would you like to log in again?
Hi-Strength 90 Spray Adhesive, 3M Industrial
Supplier: 3M
Hi-Strength 90
7000023924
89235-834CS 426.29 USD InStock
89235-834
Hi-Strength 90 Spray Adhesive, 3M Industrial
Adhesives
Spray adhesive can be used for decorative laminate bonding to tables, cabinets, and shelving, and for bonding kick plates.
  • One-minute dry time
  • High temperature resistance
  • Low misting
  • Precise spray control and variable width spray pattern
  • Clear
  • Aerosol can
Adhesive has high contact bond strength, one-minute dry time, high temperature resistance, low misting, precise spray control, and variable width spray pattern.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty