Would you like to log in again?
KLEENGUARD® A20 Breathable Particle Protection Apron, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
36550 36550
10815-827CS 296.84 USD InStock
10815-827 CA10815-827
KLEENGUARD® A20 Breathable Particle Protection Apron, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
Lightweight, stain-resistant bonded polypropylene layers provide comfort and durability.
  • Lightweight, disposable
  • Triple layer construction, breathable and pliable
  • Extra-long ties
For lab use, light assembly, and food processing areas.
28 x 40 inches
Color: white
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty