Would you like to log in again?
Mouse/ Rabbit Alkaline Phosphatase (Fast Red)
Supplier: Diagnostic BioSystems
KA50F KA50F
95025-208EA 298.1 USD InStock
95025-208 CA95025-208
Mouse/ Rabbit Alkaline Phosphatase (Fast Red)
Enzymes
This is a streptavidin biotin based detection system. Used to detect antibodies raised in mouse and rabbit.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty