Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SNCA Mouse Monoclonal Antibody [clone: 3H9]

Supplier: Rockland Immunochemicals
500-301-E56
RL500-301-E56EA 518 USD InStock
RL500-301-E56
Anti-SNCA Mouse Monoclonal Antibody [clone: 3H9]
Antibodies
Alpha Synuclein antibody is suitable for use in ELISA, Western Blotting and IF. Specific conditions should be optimized by the end user. Expect a band of approximately 15 kDa in size corresponding to the alpha synuclein Proteins in the appropriate cell lysate.

Recommended Dilutions: ELISA: 1:10000; Immunofluorescence: 1:500; Western Blot: 1:1000

Type: Primary
Antigen: SNCA
Clonality: Monoclonal
Clone: 3H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty