Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GLRA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-D65
RL600-401-D65EA 518.29 USD InStock
RL600-401-D65
Anti-GLRA1 Rabbit Polyclonal Antibody
Antibodies
Anti-Glycine Receptor (Rabbit) antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 48 kDa in size corresponding to the alpha 1 and alpha 2 subunit of the GABA(A) Receptor.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000
0.05 M Tris Chloride, 0.15 M Sodium Chloride, pH 7.4;

Type: Primary
Antigen: GLRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty