Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL17F Mouse Monoclonal Antibody [clone: 4H450.1]

Supplier: Rockland Immunochemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

209-301-B31 209-301-B31S 209-301-B61S 209-301-B61
RL209-301-B61SEA 148 USD InStock
RL209-301-B61S RL209-301-B31S RL209-301-B61 RL209-301-B31
Anti-IL17F Mouse Monoclonal Antibody [clone: 4H450.1]
Antibodies
Anti-IL-17F antibody has been tested for use in Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 0.2µg/ml;

Type: Primary
Antigen: IL17F
Clonality: Monoclonal
Clone: 4H450.1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty