Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL18 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
210-401-323S
RL210-401-323SEA 147.5 USD InStock
RL210-401-323S
Anti-IL18 Rabbit Polyclonal Antibody
Antibodies
This IgG fraction antibody of anti-Mouse IL-18 has been tested for use in neutralizations, ELISA, immunohistochem-istry and immunoblotting. Although untested, this reagent may be useful for radioimmunoassays, flow cytometry and immunoprecip-itation.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: IL18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty