Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RPE Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-387
RL600-101-387EA 605 USD InStock
RL600-101-387
Anti-RPE Goat Polyclonal Antibody
Antibodies
Suitable for ELISA, western blotting, fluorescent assays, immunoprecipitation, immunodiffusion, conjugation to magnetic particles (beads) and most other immunological methods requiring lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Tag
Antigen: RPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty