Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium ethanolate 15% (w/v) in ethanol , 99+% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium ethoxide Potassium ethylate

39161-AC 039161-AC
AA39161-ACEA 92 USD InStock
CAAA39161-AC AA39161-AC
Potassium ethanolate 15% (w/v) in ethanol , 99+% (metals basis)
Potassium ethanolate


Formula: C₂H₅KO
MW: 84.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054277
CAS Number: 917-58-8
EINECS: 213-029-0
UN: 3274
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty