Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bentonite, sodian

Supplier: Thermo Scientific Chemicals
Synonyms: Bentonite, sodium form

A15795-36 A15795-0E A15795-36 A15795-0E
AAA15795-36EA 42.98 USD InStock
CAAAA15795-0E AAA15795-36 CAAAA15795-36 AAA15795-0E
Bentonite, sodian
Bentonite, sodian
Suspending agent, gel binder, filler in ceramics, catalyst support


Formula: Al₂H₂O₁₂Si₄
MW: 360.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130611
CAS Number: 1302-78-9
EINECS: 215-108-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty