Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Cresolphthalein complexone tetrasodium salt

Supplier: Thermo Scientific Chemicals
Synonyms: o-Cresolphthalein Complexone sodium salt

A17941-06 A17941-03 A17941-06 A17941-03
AAA17941-06EA 72.79 USD InStock
CAAAA17941-06 CAAAA17941-03 AAA17941-06 AAA17941-03
o-Cresolphthalein complexone tetrasodium salt
o-Cresolphthalein complexone tetrasodium salt


Formula: C₃₂H₂₈N₂Na₄O₁₂
Storage Temperature: Ambient
MDL Number: MFCD00066371
CAS Number: 62698-54-8
EINECS: 305-335-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty