Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Hydroxycyclohexyl phenyl ketone 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (1-Hydroxycyclohexyl)(phenyl)methanone

L11377-14 L11377-06 L11377-06 L11377-14
AAAL11377-14EA 105.99 USD InStock
CAAAAL11377-14 AAAL11377-14 AAAL11377-06 CAAAAL11377-06
1-Hydroxycyclohexyl phenyl ketone 98%
1-Hydroxycyclohexyl phenyl ketone


Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Boiling Pt: >225 °C
Melting Pt: 46…49 °C
Density: 1.170
Flash Pt: >150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059561
CAS Number: 947-19-3
EINECS: 213-426-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty