Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethynyltributyltin 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethynyltri-n-butylstannaneTributylethynylstannane

L20178-06 L20178-03 L20178-03 L20178-06
AAAL20178-03EA 79.79 USD InStock
AAAL20178-03 CAAAAL20178-03 AAAL20178-06 CAAAAL20178-06
Ethynyltributyltin 96%
Ethynyltributyltin


Formula: C₁₄H₂₈Sn
MW: 315.09
Boiling Pt: 70 °C/0.2 mm
Density: 1.080
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009420
CAS Number: 994-89-8
UN: 2788
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty