Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl vinylphosphonate 97%

Supplier: Thermo Scientific Chemicals
Danger

010368-09 10368-09
AA10368-09EA 152.99 USD InStock
AA10368-09 CAAA10368-09
Diethyl vinylphosphonate 97%
Diethyl vinylphosphonate


Formula: C₆H₁₃O₃P
MW: 164.14 g/mol
Boiling Pt: 202 °C
Density: 1.068
Flash Pt: 110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009079
CAS Number: 682-30-4
EINECS: 211-663-2
UN: 3278
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty