Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium isopropoxide

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Barium i-propoxide

14523-14 14523.14
AA14523-14EA 472.99 USD InStock
AA14523-14 CAAA14523-14
Barium isopropoxide
Barium isopropoxide


Formula: C₆H₁₄BaO₂
MW: 255.501 g/mol
Melting Pt: ∼ 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00050486
CAS Number: 24363-37-9
UN: 3205
ADR: 4.2,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty