Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium silicon oxide

Supplier: Thermo Scientific Chemicals
Warning

15122-36 15122.36
AA15122-36EA 229.99 USD InStock
AA15122-36 CAAA15122-36
Barium silicon oxide
Barium silicon oxide


Formula: BaO and SiO₂
MW: 213.40 g/mol
Melting Pt: 1604 °C
Density: 4,399
Storage Temperature: Ambient
MDL Number: MFCD00014186
CAS Number: 13255-26-0
UN: 1564
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty