Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(pentamethylcyclopentadienyl)titanium(IV) dichloride

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bis(pentamethylcyclopentadienyl)titanium dichloride

39570-06 39570-06 039570-03 39570-03
AA39570-06EA 810.23 USD InStock
CAAA39570-03 AA39570-06 CAAA39570-06 AA39570-03
Bis(pentamethylcyclopentadienyl)titanium(IV) dichloride
Bis(pentamethylcyclopentadienyl)titanium(IV) dichloride


Formula: C₂₀H₃₀Cl₂Ti
MW: 389.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049153
CAS Number: 11136-36-0
UN: 3261
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty