Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silicon(IV) nitride, MgO binder, sputtering target ≥99.9% (metals basis), Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

41090-KS 041090-KS
AA41090-KSEA 1422.99 USD InStock
AA41090-KS CAAA41090-KS
Silicon(IV) nitride, MgO binder, sputtering target ≥99.9% (metals basis), Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)
Silicon(IV) nitride, MgO binder, sputtering target


Formula: Si₃N₄
Storage Temperature: Ambient
MDL Number: MFCD00011230
EINECS: 234-796-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty