Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbon conductive cement adhesive

Supplier: Thermo Scientific Chemicals
Danger

41212-15 41212-15
AA41212-15EA 173.99 USD InStock
CAAA41212-15 AA41212-15
Carbon conductive cement adhesive
Carbon conductive cement adhesive
  • Form: Paste


Storage Temperature: Ambient
MDL Number: MFCD00144065
UN: 1133
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty