Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polystyrene standard, M.W. 1,300, Mw/Mn 1.06, Thermo Scientific

Supplier: Thermo Scientific
041922-03 41922-03
AA41922-03EA 104.15 USD InStock
CAAA41922-03 AA41922-03
Polystyrene standard, M.W. 1,300, Mw/Mn 1.06, Thermo Scientific
Standards Organic Standards
Uses: Molecular weight standard for gel permeation chromatography (GPC)

CAS No.: 9003-53-6
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD00084450

Applications: Molecular weight standard for gel permeation chromatography (GPC)

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty