Would you like to log in again?
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Diameter 2 mm (0.08 in) Type 316
Supplier: Thermo Scientific Chemicals
042019-KM 042019-KN 42019-KM 42019-KN
AA42019-KMEA 34.19 USD InStock
AA42019-KM AA42019-KN CAAA42019-KN CAAA42019-KM
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Diameter 2 mm (0.08 in) Type 316
Stainless steel
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty