Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium(IV) solution 0.25 N in dilute nitric acid etchant solution

Supplier: Thermo Scientific Chemicals
Danger

42110-K7 042110-K7
AA42110-K7EA 123.99 USD InStock
CAAA42110-K7 AA42110-K7
Cerium(IV) solution 0.25 N in dilute nitric acid etchant solution
Cerium(IV) solution


Storage Temperature: Ambient
MDL Number: MFCD01866052
UN: 3098
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty