Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane-[D₁₂] (98% D) (Isotopic)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: n-Pentane-[²H₁₂]

42340.03 42340-03
AA42340-03EA 155.99 USD InStock
CAAA42340-03 AA42340-03
n-Pentane-[D₁₂] (98% D) (Isotopic)
n-Pentane-[D12]


Formula: C₅D₁₂
MW: 84.22
Boiling Pt: 36 °C
Melting Pt: –130 °C
Density: 0.731
Flash Pt: –49 °C (–56 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00190495
CAS Number: 2031-90-5
EINECS: 203-692-4
UN: 1265
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty