Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdenite naturally occurring mineral, grains ∼ 0.06-0.19 in

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: Molybdenum(IV) sulphideMolybdenum(IV) sulfide

42642-06 042642-14 42642-03 42642-14 042642-06 042642-03
AA42642-06EA 75.09 USD InStock
AA42642-06 CAAA42642-03 CAAA42642-14 AA42642-14 AA42642-03 CAAA42642-06
Molybdenite naturally occurring mineral, grains ∼ 0.06-0.19 in
Molybdenite
For grain mounts, XRD and microprobe standards, identification of unknown minerals
Hardness 1-1.5; Sp.Gr. 4.62-4.73
Soluble in sulfuric and nitric acid


Formula: MoS₂
MW: 160.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003470
CAS Number: 1309-56-4
EINECS: 215-172-4
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty