Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver conductive adhesive paste

Supplier: Thermo Scientific Chemicals
Warning

44075-18 44075-18
AA44075-18EA 611.99 USD InStock
AA44075-18 CAAA44075-18
Silver conductive adhesive paste
Silver conductive adhesive paste
Temperature stability range -65° to 275°C


Storage Temperature: Ambient
MDL Number: MFCD01868500
EINECS: 232-033-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty