Would you like to log in again?
Brass (alloy 260), foil, Thickness 1.3 mm (0.05 in)
Supplier: Thermo Scientific Chemicals
45234-GT 45234-HB
AA45234-HBEA 83.99 USD InStock
AA45234-HB AA45234-GT
Brass (alloy 260), foil, Thickness 1.3 mm (0.05 in)
Brass (alloy 260)
Typical composition of alloy: Cu 68.5-71.5%, Pb 0.07% max, Fe 0.05% max, Zn remainder
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty