Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gallium arsenide, Optical Window, Diameter 12.7 mm (0.5 in), Thickness 1 mm (0.04 in)

Supplier: Thermo Scientific Chemicals
Danger

045628-KS
AA45628-KSEA 281.99 USD InStock
AA45628-KS
Gallium arsenide, Optical Window, Diameter 12.7 mm (0.5 in), Thickness 1 mm (0.04 in)
Gallium arsenide


Formula: AsGa
MW: 144.64 g/mol
Melting Pt: 1238 °C
Density: 5.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011017
CAS Number: 1303-00-0
EINECS: 215-114-8
UN: 1557
ADR: 6.1,II
Merck Index: 12,04365
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty