Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(II) pyrophosphate

Supplier: Thermo Scientific Chemicals
Warning

089785-30 89785-18 89785-30 089785-18
AA89785-18EA 49.29 USD InStock
AA89785-18 AA89785-30 CAAA89785-30 CAAA89785-18
Tin(II) pyrophosphate
Tin(II) pyrophosphate
Insoluble in water


Formula: O₇P₂Sn₂
Melting Pt: 400 °C (dec.)
Density: 4,009
Storage Temperature: Ambient
MDL Number: MFCD00049544
CAS Number: 15578-26-4
EINECS: 239-635-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty