Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Fluorobenzo-2-oxa-13-diazole-4-sulfonic acid ammonium salt 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonateAmmonium 7-fluoro-2,1,3-benzoxadiazole-4-sulfonate

B21767-MF B21767-MD B21767-MD B21767-MF
AAB21767-MDEA 2315.62 USD InStock
CAAAB21767-MD AAB21767-MD AAB21767-MF CAAAB21767-MF
7-Fluorobenzo-2-oxa-13-diazole-4-sulfonic acid ammonium salt 99%
7-Fluorobenzo-2-oxa-13-diazole-4-sulfonic acid ammonium salt


Formula: C₆H₆FN₃O₄S
MW: 235.189 g/mol
Melting Pt: ca. 285…295°C dec. (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02682109
CAS Number: 84806-27-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty