Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoropropionic acid 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Fluoropropionicacid

H25841-03 H25841-MD
AAH25841-MDEA 94.88 USD InStock
AAH25841-MD AAH25841-03
2-Fluoropropionic acid 96%
2-Fluoropropionic acid


Formula: C₃H₅FO₂
MW: 92.07 g/mol
Boiling Pt: 66…67 °C/30 mm
Density: 1.214
Flash Pt: 82 °C(180 °F)
Storage Temperature: Ambient
MDL Number: MFCD06247714
CAS Number: 6087-13-4
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty