Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-HomoArg-OH.HClH-HoArg-OH·HClL(+)-Homoarginine hydrochlorideH-HomoArg-OH·HCl

H27387-14 H27387-03 H27387-06
AAH27387-06EA 257.99 USD InStock
AAH27387-06 AAH27387-14 AAH27387-03
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥98%
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride


Formula: C₇H₁₆N₄O₂·HCl
MW: 224.7 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012622
CAS Number: 1483-01-8
EINECS: 216-045-6
Order Now
Specification Test Results
Optical rotation +18° (c=1 in 1N HCl)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty