Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-1-methylcyclopropyl ketone 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-(1-Methylcyclopropyl)ethanoneMethyl 1-methylcyclopropyl ketone

L05394-06 L05394-06
AAAL05394-06EA 99.59 USD InStock
CAAAAL05394-06 AAAL05394-06
Methyl-1-methylcyclopropyl ketone 96%
Methyl-1-methylcyclopropyl ketone


Formula: C₆H₁₀O
MW: 98.145 g/mol
Boiling Pt: 126…128 °C
Density: 0.896
Flash Pt: 23 °C(73 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010245
CAS Number: 1567-75-5
EINECS: 216-369-8
UN: 1224
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty