Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-1-boc-piperidine-4-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Amino-1-(tert-butoxycarbonyl)piperidine-4-carboxylic acid

L19794-MD L19794-03 L19794-03
AAAL19794-03EA 180.99 USD InStock
AAAL19794-03 CAAAAL19794-MD CAAAAL19794-03
4-Amino-1-boc-piperidine-4-carboxylic acid 98%
4-Amino-1-Boc-piperidine-4-carboxylic acid


Formula: C₁₁H₂₀N₂O₄
MW: 244.29 g/mol
Melting Pt: ca 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01318728
CAS Number: 183673-71-4
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty