Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(cyclopentadienyl)manganese, sublimated

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Manganocene

ampouled under argon

14535-06 014535-06
AA14535-06EA 748.99 USD InStock
AA14535-06 CAAA14535-06
Bis(cyclopentadienyl)manganese, sublimated
Bis(cyclopentadienyl)manganese


Formula: C₁₀H₁₀Mn
MW: 185.13 g/mol
Boiling Pt: 245 °C
Melting Pt: 175 °C
Flash Pt: 52 °C (126 °F)
Storage Temperature: Ambient
MDL Number: MFCD00045468
CAS Number: 1271-27-8
EINECS: 215-038-5
UN: 3181
ADR: 4.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty