Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium carbonate standardised solution 0.05 N in aqueous solution

Supplier: Thermo Scientific Chemicals
Synonyms: Bisodium carbonateCarbonic acid, disodium saltCarbonic acid sodium saltDisodium carbonateNa-XSodaSoda ash

35609-K2 035609-K2
AA35609-K2EA 61.19 USD InStock
CAAA35609-K2 AA35609-K2
Sodium carbonate standardised solution 0.05 N in aqueous solution
Sodium carbonate


Formula: NaCO₃
MW: 105.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/89/16640589.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty