Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylaluminium dichloride 1 M in hexanes

Supplier: Thermo Scientific Chemicals
Danger

041712-AK 041712-AB 41712-AB 41712-AK
AA41712-AKEA 132.99 USD InStock
CAAA41712-AK AA41712-AK CAAA41712-AB AA41712-AB
Ethylaluminium dichloride 1 M in hexanes
Ethylaluminium dichloride


Formula: C₂H₅AlCl₂
Boiling Pt: 68…70 °C
Density: 0,729
Flash Pt: –22 °C (–9 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000457
CAS Number: 563-43-9
EINECS: 209-248-6
UN: 3394
ADR: 4.2,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty