Would you like to log in again?
Turbine-Type Large Scale Agitator, PTFE, 28 mm, Ace Glass Incorporated
Supplier: Ace Glass
8093-25 8093-35
89062-174EA 1268.98 USD InStock
89062-174 89062-176
Turbine-Type Large Scale Agitator, PTFE, 28 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
A four-blade, 45° pitch, turbine-type agitator designed for use at the upper section of 28mm stirring shafts (see 89062-030 series)
The PTFE agitator is supplied with PTFE bolt and nut for securing to the shaft to prevent detachment during operation. Blades are 6.35mm (¼") thick. The increased length and depth of the blades imparts more energy to the load. Agitator is not supplied with shaft.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty