Would you like to log in again?
Air Sampling Sweep Elbow, Ace Glass Incorporated
Supplier: Ace Glass
7490-37
89059-172EA 336.68 USD InStock
89059-172
Air Sampling Sweep Elbow, Ace Glass Incorporated
Fittings and Connectors
Sweep elbow features #25 Ace-Thred connections on each end
Can be used with a #25 Ace-Thred bushing such as 89059-226, or with couplings such as 89052-644. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty