Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allyl acetate ≥98.0%

Supplier: TCI America
Synonyms: Acetic acid allyl ester

A0020-500ML A0020-25ML
TCA0020-25MLEA 46.54 USD InStock
TCA0020-25ML TCA0020-500ML
Allyl acetate ≥98.0%
Allyl acetate
CAS Number: 591-87-7
MDL Number: MFCD00008721
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 15
Freezing point (°C): -96
Specific Gravity (20/20): 0.93
Storage Temperature: 0-10°C

Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acetic Acid Allyl Ester


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 105 °C
Melting Pt: –96 °C
Density: 0.93 (20 °C)
Flash Pt: 15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008721
CAS Number: 591-87-7
EINECS: 209-734-8
UN: 2333
ADR: 3,II
Merck Index: 12,00293
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/58/28927458.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty