Synonyms:
Acetic acid amyl ester
, n-Pentyl acetate
, Pentyl acetate
, Acetic acid pentyl ester
A0021-25ML
A0021-500ML
TCA0021-25MLEA
28.12
USD
InStock
TCA0021-25ML
TCA0021-500ML
Amyl acetate ≥99.0%
Amyl acetate
CAS Number: 628-63-7
MDL Number: MFCD00009500
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 45
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: Acetic Acid Amyl Ester, Pentyl Acetate