Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.5%

Supplier: TCI America
Danger
Synonyms: Acetic acid ethyl esterAcetic etherAcetidinAcetoxyethaneEthyl acetic esterEthyl ethanoateVinegar naphtha

A0030-250ML
TCA0030-250MLEA 50.93 USD InStock
TCA0030-250ML
Ethyl acetate ≥99.5%
Ethyl acetate
[for Spectrophotometry]
CAS Number: 141-78-6
MDL Number: MFCD00009171
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: 99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 77
Flash Point (°C): -3
Specific Gravity (20/20): 0.90

Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Acetic Acid Ethyl Ester


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C
Melting Pt: –83 °C
Density: 0.90 (20 °C)
Flash Pt: –3 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/69/28927469.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.5% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty